siRNA Details
| Gene Symbol: Ttyh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_221081.8 | XP_221081.7 | 4 | 684 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttyh2| Gene Symbol: Ttyh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_221081.8 | XP_221081.7 | 4 | 684 | ||