siRNA Details
| Gene Symbol: Nup85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025401.1 | NP_001020572.1 | 8 | 666 | ||
| GenBank mRNA | BC098031.1 | - | 8 | 666 | 7456325 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nup85| Gene Symbol: Nup85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025401.1 | NP_001020572.1 | 8 | 666 | ||
| GenBank mRNA | BC098031.1 | - | 8 | 666 | 7456325 | |