siRNA Details
| Gene Symbol: Pycr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105857.2 | NP_001099327.1 | 3 | 244 | ||
| XM_008768472.1 | XP_008766694.1 | 4 | 441 | |||
| XM_008768473.1 | XP_008766695.1 | 4 | 375 | |||
| XM_008768474.1 | XP_008766696.1 | 4 | 365 | |||
| XM_008768475.1 | XP_008766697.1 | 4 | 441 | |||

Pycr1
Genomic Map