siRNA Details
| Gene Symbol: St6galnac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105859.1 | NP_001099329.1 | 2 | 407 | ||
| GenBank mRNA | AJ634458.1 | - | 2 | 306 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

St6galnac1| Gene Symbol: St6galnac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105859.1 | NP_001099329.1 | 2 | 407 | ||
| GenBank mRNA | AJ634458.1 | - | 2 | 306 | ||