siRNA Details
Gene Symbol: Yars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009627.1 | NP_001009627.1 | 1 | 634 | ||
GenBank mRNA | BC088224.1 | - | 1 | 634 | 7322099 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Yars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009627.1 | NP_001009627.1 | 1 | 634 | ||
GenBank mRNA | BC088224.1 | - | 1 | 634 | 7322099 |