siRNA Details
Gene Symbol: Pkp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100499.1 | NP_001093969.1 | 1456 | |||
GenBank mRNA | BC092602.1 | - | 1, 2 | 63 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pkp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100499.1 | NP_001093969.1 | 1456 | |||
GenBank mRNA | BC092602.1 | - | 1, 2 | 63 |