siRNA Details
| Gene Symbol: Top3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105861.1 | NP_001099331.1 | 14 | 1710 | ||
| XM_006248671.2 | XP_006248733.1 | 13 | 1844 | |||
| XM_006248673.2 | XP_006248735.1 | 13 | 1656 | |||
| GenBank mRNA | FQ212059.1 | - | 13 | 1665 | ||

Top3b
Genomic Map