siRNA Details
| Gene Symbol: Trmt2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011895.1 | NP_001011895.1 | 8, 9 | 1574 | ||
| XM_006248604.1 | XP_006248666.1 | 8, 9 | 1786 | |||
| XM_006248605.1 | XP_006248667.1 | 8, 9 | 1545 | |||
| GenBank mRNA | BC083619.1 | - | 8, 9 | 1574 | 7133938 | |

Trmt2a
Genomic Map