siRNA Details
Gene Symbol: Cdc45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105866.2 | NP_001099336.1 | 1, 2 | 66 | ||
XM_008768866.1 | XP_008767088.1 | 2, 3 | 388 | |||
GenBank mRNA | BC161946.1 | - | 1, 2 | 66 | 9025372 | |
FQ232109.1 | - | 1, 2 | 68 |
