siRNA Details
| Gene Symbol: Cdc45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105866.2 | NP_001099336.1 | 17, 18 | 1653 | ||
| XM_008768866.1 | XP_008767088.1 | 18, 19 | 1975 | |||
| GenBank mRNA | BC161946.1 | - | 17, 18 | 1653 | 9025372 | |

Cdc45
Genomic Map