siRNA Details
Gene Symbol: Rfc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105869.2 | NP_001099339.1 | 7 | 668 | ||
GenBank mRNA | BC168166.1 | - | 7 | 668 | 8374426 | |
FQ228232.1 | - | 7 | 700 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rfc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105869.2 | NP_001099339.1 | 7 | 668 | ||
GenBank mRNA | BC168166.1 | - | 7 | 668 | 8374426 | |
FQ228232.1 | - | 7 | 700 |