siRNA Details
| Gene Symbol: P3h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025627.1 | NP_001020798.1 | 5 | 1127 | ||
| GenBank mRNA | BC099107.1 | - | 5 | 1127 | 7460856 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

P3h2| Gene Symbol: P3h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025627.1 | NP_001020798.1 | 5 | 1127 | ||
| GenBank mRNA | BC099107.1 | - | 5 | 1127 | 7460856 | |