siRNA Details
| Gene Symbol: Hrasls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105871.1 | NP_001099341.1 | 4 | 590 | ||
| XM_006248510.2 | XP_006248572.1 | 4 | 569 | |||
| GenBank mRNA | AB510983.1 | - | 3 | 440 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hrasls| Gene Symbol: Hrasls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105871.1 | NP_001099341.1 | 4 | 590 | ||
| XM_006248510.2 | XP_006248572.1 | 4 | 569 | |||
| GenBank mRNA | AB510983.1 | - | 3 | 440 | ||