siRNA Details
| Gene Symbol: Atp13a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191658.2 | NP_001178587.2 | 25 | 2793 | ||
| XM_006248511.2 | XP_006248573.1 | 24 | 2567 | |||
| XM_006248512.2 | XP_006248574.1 | 25 | 3232 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp13a4| Gene Symbol: Atp13a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191658.2 | NP_001178587.2 | 25 | 2793 | ||
| XM_006248511.2 | XP_006248573.1 | 24 | 2567 | |||
| XM_006248512.2 | XP_006248574.1 | 25 | 3232 | |||