siRNA Details
| Gene Symbol: Atp13a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191658.2 | NP_001178587.2 | 20 | 2409 | ||
| XM_006248511.2 | XP_006248573.1 | 19 | 2183 | |||
| XM_006248512.2 | XP_006248574.1 | 20 | 2848 | |||
| XM_008768796.1 | XP_008767018.1 | 20 | 2850 | |||

Atp13a4
Genomic Map