siRNA Details
Gene Symbol: Lsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013421.1 | NP_001013439.1 | 9, 10 | 1275 | ||
GenBank mRNA | BC091338.1 | - | 9, 10 | 1275 | 7315079 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013421.1 | NP_001013439.1 | 9, 10 | 1275 | ||
GenBank mRNA | BC091338.1 | - | 9, 10 | 1275 | 7315079 |