siRNA Details
| Gene Symbol: Mylk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105874.2 | NP_001099344.2 | 8 | 905 | ||
| XM_006248433.2 | XP_006248495.1 | 8 | 1213 | |||
| XM_006248434.2 | XP_006248496.1 | 8 | 995 | |||
| XM_008768711.1 | XP_008766933.1 | 8 | 1214 | |||

Mylk
Genomic Map