siRNA Details
| Gene Symbol: Polq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105878.1 | NP_001099348.1 | 17 | 3043 | ||
| XM_006248390.2 | XP_006248452.1 | 14 | 3043 | |||
| XM_006248391.2 | XP_006248453.1 | 14 | 3452 | |||
| XM_008768716.1 | XP_008766938.1 | 16 | 3066 | |||

Polq
Genomic Map