siRNA Details
| Gene Symbol: Stxbp5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271250.1 | NP_001258179.1 | 14 | 1607 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stxbp5l| Gene Symbol: Stxbp5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271250.1 | NP_001258179.1 | 14 | 1607 | ||