siRNA Details
| Gene Symbol: Poglut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100652.1 | NP_001094122.1 | 7 | 754 | ||
| GenBank mRNA | BC091408.1 | - | 6 | 526 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Poglut1| Gene Symbol: Poglut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100652.1 | NP_001094122.1 | 7 | 754 | ||
| GenBank mRNA | BC091408.1 | - | 6 | 526 | ||