siRNA Details
| Gene Symbol: Tmem39a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013865.1 | NP_001013887.1 | 9 | 1633 | ||
| XM_008768717.1 | XP_008766939.1 | 8 | 1588 | |||
| GenBank mRNA | BC085845.1 | - | 9 | 1633 | 7188963 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem39a| Gene Symbol: Tmem39a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013865.1 | NP_001013887.1 | 9 | 1633 | ||
| XM_008768717.1 | XP_008766939.1 | 8 | 1588 | |||
| GenBank mRNA | BC085845.1 | - | 9 | 1633 | 7188963 | |