siRNA Details
| Gene Symbol: Slc9c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008762.2 | NP_001008762.2 | 12 | 1617 | ||
| XM_008768722.1 | XP_008766944.1 | 11 | 2050 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc9c1| Gene Symbol: Slc9c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008762.2 | NP_001008762.2 | 12 | 1617 | ||
| XM_008768722.1 | XP_008766944.1 | 11 | 2050 | |||