siRNA Details
| Gene Symbol: Hoxd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271038.1 | NP_001257967.1 | 3 | 1065 | ||
| XM_008761880.1 | XP_008760102.1 | 2 | 1325 | |||
| XM_008761881.1 | XP_008760103.1 | 3 | 803 | |||
| GenBank mRNA | M37569.1 | - | 1 | 135 | ||

Hoxd3
Genomic Map