siRNA Details
| Gene Symbol: B3galt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105887.1 | NP_001099357.1 | 3 | 1084 | ||
| XM_008768569.1 | XP_008766791.1 | 3 | 3139 | |||
| XM_008768570.1 | XP_008766792.1 | 3 | 1236 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B3galt5| Gene Symbol: B3galt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105887.1 | NP_001099357.1 | 3 | 1084 | ||
| XM_008768569.1 | XP_008766791.1 | 3 | 3139 | |||
| XM_008768570.1 | XP_008766792.1 | 3 | 1236 | |||