siRNA Details
| Gene Symbol: Senp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105888.1 | NP_001099358.1 | 11 | 1645 | ||
| XM_006248244.2 | XP_006248306.1 | 11 | 2600 | |||
| XM_006248247.2 | XP_006248309.1 | 10 | 2403 | |||
| XM_006248248.2 | XP_006248310.1 | 9 | 2210 | |||

Senp7
Genomic Map