siRNA Details
| Gene Symbol: Tmem30c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105889.1 | NP_001099359.1 | 6 | 636 | ||
| XM_008768632.1 | XP_008766854.1 | 6 | 873 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem30c| Gene Symbol: Tmem30c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105889.1 | NP_001099359.1 | 6 | 636 | ||
| XM_008768632.1 | XP_008766854.1 | 6 | 873 | |||