siRNA Details
| Gene Symbol: Cldnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006955.1 | NP_001006956.1 | 2, 3 | 391 | ||
| XM_006248204.2 | XP_006248266.1 | 3, 4 | 583 | |||
| XM_006248205.2 | XP_006248267.1 | 2, 3 | 532 | |||
| XM_006248206.2 | XP_006248268.1 | 3, 4 | 434 | |||
| XM_008768633.1 | XP_008766855.1 | 2, 3 | 374 | |||
| GenBank mRNA | BC083649.1 | - | 2, 3 | 391 | 7135100 | |

Cldnd1
Genomic Map