siRNA Details
| Gene Symbol: Hlcs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248111.2 | XP_006248173.1 | 4 | 1552 | ||
| XM_006248112.2 | XP_006248174.1 | 4 | 1619 | |||
| XM_006248114.2 | XP_006248176.1 | 3 | 1413 | |||
| XM_008768611.1 | XP_008766833.1 | 4 | 751 | |||
| XM_008768612.1 | XP_008766834.1 | 3 | 616 | |||
| XM_221630.8 | XP_221630.6 | 3 | 732 | |||

Hlcs
Genomic Map