siRNA Details
| Gene Symbol: Donson | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008287.1 | NP_001008288.1 | 1, 2 | 422 | ||
| XM_006248047.2 | XP_006248109.1 | 1, 2 | 416 | |||
| XM_006248048.2 | XP_006248110.1 | 1, 2 | 416 | |||
| GenBank mRNA | BC085847.1 | - | 1, 2 | 422 | 7189203 | |

Donson
Genomic Map