siRNA Details
| Gene Symbol: Ifnar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105893.1 | NP_001099363.2 | 8 | 1342 | ||
| NM_001271316.1 | NP_001258245.1 | 9 | 1532 | |||
| GenBank mRNA | FQ210936.1 | - | 9 | 1332 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifnar1| Gene Symbol: Ifnar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105893.1 | NP_001099363.2 | 8 | 1342 | ||
| NM_001271316.1 | NP_001258245.1 | 9 | 1532 | |||
| GenBank mRNA | FQ210936.1 | - | 9 | 1332 | ||