siRNA Details
| Gene Symbol: Ncam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203409.1 | NP_981954.1 | 15 | 2155 | ||
| GenBank mRNA | AY495695.1 | - | 15 | 2155 | ||
| AY495696.1 | - | 15 | 2275 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ncam2| Gene Symbol: Ncam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203409.1 | NP_981954.1 | 15 | 2155 | ||
| GenBank mRNA | AY495695.1 | - | 15 | 2155 | ||
| AY495696.1 | - | 15 | 2275 | |||