siRNA Details
Gene Symbol: Tmprss15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105895.1 | NP_001099365.1 | 26 | 3005 | ||
XM_008768527.1 | XP_008766749.1 | 24 | 2819 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmprss15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105895.1 | NP_001099365.1 | 26 | 3005 | ||
XM_008768527.1 | XP_008766749.1 | 24 | 2819 |