siRNA Details
| Gene Symbol: Cct8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105897.2 | NP_001099367.2 | 14 | 1651 | ||
| GenBank mRNA | DY319674.1 | - | 4 | 344 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cct8| Gene Symbol: Cct8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105897.2 | NP_001099367.2 | 14 | 1651 | ||
| GenBank mRNA | DY319674.1 | - | 4 | 344 | ||