siRNA Details
| Gene Symbol: Rbm11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105898.1 | NP_001099368.1 | 4 | 466 | ||
| XM_006247994.2 | XP_006248056.1 | 4 | 507 | |||
| XM_006247995.2 | XP_006248057.1 | 4 | 507 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm11| Gene Symbol: Rbm11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105898.1 | NP_001099368.1 | 4 | 466 | ||
| XM_006247994.2 | XP_006248056.1 | 4 | 507 | |||
| XM_006247995.2 | XP_006248057.1 | 4 | 507 | |||