siRNA Details
| Gene Symbol: Cggbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105900.1 | NP_001099370.1 | 3 | 658 | ||
| XM_006247975.2 | XP_006248037.1 | 3 | 715 | |||
| XM_008768528.1 | XP_008766750.1 | 2 | 982 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cggbp1| Gene Symbol: Cggbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105900.1 | NP_001099370.1 | 3 | 658 | ||
| XM_006247975.2 | XP_006248037.1 | 3 | 715 | |||
| XM_008768528.1 | XP_008766750.1 | 2 | 982 | |||