siRNA Details
| Gene Symbol: Mcoln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105903.1 | NP_001099373.1 | 13 | 1752 | ||
| XM_006248754.2 | XP_006248816.1 | 14 | 2324 | |||
| XM_006248755.2 | XP_006248817.1 | 14 | 2278 | |||
| XM_006248756.2 | XP_006248818.1 | 13 | 2247 | |||

Mcoln1
Genomic Map