siRNA Details
| Gene Symbol: Hsph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011901.1 | NP_001011901.1 | 7 | 890 | ||
| XM_008768947.1 | XP_008767169.1 | 6 | 651 | |||
| GenBank mRNA | BC081945.1 | - | 7 | 890 | 7121434 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hsph1| Gene Symbol: Hsph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011901.1 | NP_001011901.1 | 7 | 890 | ||
| XM_008768947.1 | XP_008767169.1 | 6 | 651 | |||
| GenBank mRNA | BC081945.1 | - | 7 | 890 | 7121434 | |