siRNA Details
| Gene Symbol: Nptx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034199.1 | NP_001029371.1 | 5 | 1317 | ||
| GenBank mRNA | CO388671.1 | - | 1 | 65 | ||
| S82649.1 | - | 1251 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nptx2| Gene Symbol: Nptx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034199.1 | NP_001029371.1 | 5 | 1317 | ||
| GenBank mRNA | CO388671.1 | - | 1 | 65 | ||
| S82649.1 | - | 1251 | ||||