siRNA Details
| Gene Symbol: Rbak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191664.1 | NP_001178593.1 | 3 | 730 | ||
| XM_006248867.1 | XP_006248929.1 | 5 | 1029 | |||
| XM_006248868.1 | XP_006248930.1 | 5 | 1026 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbak| Gene Symbol: Rbak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191664.1 | NP_001178593.1 | 3 | 730 | ||
| XM_006248867.1 | XP_006248929.1 | 5 | 1029 | |||
| XM_006248868.1 | XP_006248930.1 | 5 | 1026 | |||