siRNA Details
| Gene Symbol: Snx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105912.1 | NP_001099382.1 | 6 | 685 | ||
| XM_006248920.2 | XP_006248982.1 | 6 | 680 | |||
| XM_006248921.2 | XP_006248983.1 | 6 | 1080 | |||
| XM_006248923.2 | XP_006248985.1 | 6 | 953 | |||

Snx8
Genomic Map