siRNA Details
| Gene Symbol: Micall2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248975.2 | XP_006249037.1 | 13 | 2855 | ||
| GenBank mRNA | FQ232177.1 | - | 7 | 682 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Micall2| Gene Symbol: Micall2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248975.2 | XP_006249037.1 | 13 | 2855 | ||
| GenBank mRNA | FQ232177.1 | - | 7 | 682 | ||