siRNA Details
| Gene Symbol: Get4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163320.1 | NP_001156792.1 | 8 | 987 | ||
| XM_006248927.2 | XP_006248989.1 | 8 | 3223 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Get4| Gene Symbol: Get4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163320.1 | NP_001156792.1 | 8 | 987 | ||
| XM_006248927.2 | XP_006248989.1 | 8 | 3223 | |||