siRNA Details
| Gene Symbol: Gjc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769070.1 | XP_008767292.1 | 2 | 777 | ||
| XM_008769071.1 | XP_008767293.1 | 2 | 585 | |||
| XM_221997.7 | XP_221997.5 | 2 | 1753 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gjc3| Gene Symbol: Gjc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769070.1 | XP_008767292.1 | 2 | 777 | ||
| XM_008769071.1 | XP_008767293.1 | 2 | 585 | |||
| XM_221997.7 | XP_221997.5 | 2 | 1753 | |||