siRNA Details
Gene Symbol: Gjc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769070.1 | XP_008767292.1 | 2 | 1057 | ||
XM_008769071.1 | XP_008767293.1 | 2 | 865 | |||
XM_221997.7 | XP_221997.5 | 2 | 2033 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gjc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769070.1 | XP_008767292.1 | 2 | 1057 | ||
XM_008769071.1 | XP_008767293.1 | 2 | 865 | |||
XM_221997.7 | XP_221997.5 | 2 | 2033 |