siRNA Details
| Gene Symbol: Mcm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004203.3 | NP_001004203.3 | 3 | 244 | ||
| XM_006249005.2 | XP_006249067.1 | 3 | 366 | |||
| XM_006249006.2 | XP_006249068.1 | 3 | 249 | |||
| GenBank mRNA | BC078973.1 | - | 3 | 244 | 7111847 | |
| DQ278199.1 | - | 3 | 292 | |||
| DQ278200.1 | - | 837 | ||||

Mcm7
Genomic Map