siRNA Details
| Gene Symbol: Actl6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105917.1 | NP_001099387.2 | 12 | 1162 | ||
| XM_006249120.2 | XP_006249182.1 | 12 | 1243 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Actl6b| Gene Symbol: Actl6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105917.1 | NP_001099387.2 | 12 | 1162 | ||
| XM_006249120.2 | XP_006249182.1 | 12 | 1243 | |||