siRNA Details
| Gene Symbol: Fkbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105922.1 | NP_001099392.1 | 2 | 145 | ||
| XM_006249131.2 | XP_006249193.1 | 2 | 164 | |||
| XM_008769130.1 | XP_008767352.1 | 2 | 164 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fkbp6| Gene Symbol: Fkbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105922.1 | NP_001099392.1 | 2 | 145 | ||
| XM_006249131.2 | XP_006249193.1 | 2 | 164 | |||
| XM_008769130.1 | XP_008767352.1 | 2 | 164 | |||