siRNA Details
| Gene Symbol: Cct6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033684.1 | NP_001028856.1 | 10 | 1134 | ||
| GenBank mRNA | BC091282.1 | - | 4 | 256 | ||
| BC104703.1 | - | 10 | 1134 | 7300674 | ||
| FQ216833.1 | - | 10 | 1152 | |||
| FQ218432.1 | - | 10 | 1181 | |||

Cct6a
Genomic Map