siRNA Details
| Gene Symbol: Zcchc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105929.1 | NP_001099399.1 | 8 | 831 | ||
| XM_006249333.2 | XP_006249395.1 | 7 | 805 | |||
| XM_006249334.2 | XP_006249396.1 | 7 | 805 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zcchc8| Gene Symbol: Zcchc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105929.1 | NP_001099399.1 | 8 | 831 | ||
| XM_006249333.2 | XP_006249395.1 | 7 | 805 | |||
| XM_006249334.2 | XP_006249396.1 | 7 | 805 | |||