siRNA Details
| Gene Symbol: Zcchc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105929.1 | NP_001099399.1 | 11 | 1196 | ||
| XM_006249333.2 | XP_006249395.1 | 10 | 1170 | |||
| XM_006249334.2 | XP_006249396.1 | 10 | 1170 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zcchc8| Gene Symbol: Zcchc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105929.1 | NP_001099399.1 | 11 | 1196 | ||
| XM_006249333.2 | XP_006249395.1 | 10 | 1170 | |||
| XM_006249334.2 | XP_006249396.1 | 10 | 1170 | |||